Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HeLa, Mouse liver, Mouse kidney. The target is usually found in the following locations: Membrane, Single-pass type II membrane protein.
Immunogen sequence: IGHGNRMHHH EHHHLQAPNK EDILKISEDE RMELSKSFRV YCIILVKPKD VSLWAAVKET WTKHCDKAEF FSSENVKVFE SINMDTNDMW LMMRKAYKYA FDKYRDQYNW FFLARPTTFA IIENLKYFLL KKDPSQPFYL GHTIKSGDLE YVGMEGGIVL S
This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1Gal-T2; C1GALT1-specific chaperone 1; C38H2-L1; C38H2-like protein 1; Core 1 beta1,3-galactosyltransferase 2; core 1 beta3-Gal-T2; Core 1 beta3-galactosyltransferase-specific molecular chaperone; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 2; mC1Gal-T2
Gene Aliases: 1500002I11Rik; AB030184; C1GALT1C1; C1GALT2; C38H2-L1; C81205; COSMC; HSPC067; MST143; MSTP143; RGD1311230; TNPS; UNQ273/PRO310
UniProt ID: (Human) Q8WWS3, (Mouse) Q9JMG2, (Rat) Q499P3
Entrez Gene ID: (Human) 29071, (Mouse) 59048, (Rat) 302499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support