Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLPLLRCVP RVLGSSVAGL RAAAPASPFR QLLQPAPRLC TRPFGLLSVR AGSERRPGLL RPRGPCACGC GCGSLHTDGD KAFVDFLSDE IKEERKIQKH KTLPKMSGGW ELELNGTEAK LVRKVAGEKI TVTFNINNSI PPTFDGEEEP SQGQKVEEQE PELTSTPNFV VEVIKNDDGK KALVLDCHYP EDEVGQEDEA ESDIFSIREV SFQSTGESEW KDTNYTLNTD SLDWALYDHL MDFLADRGVD NTFADELVEL STALEHQEYI TFLEDLKSFV KSQ (1-282 aa encoded by BC013731)
Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1qBP; Complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; GC1q-R protein; Glycoprotein gC1qBP; Hyaluronan-binding protein 1; Mitochondrial matrix protein p32; p33; RPA; SF2AP32; splicing factor SF2-associated protein
Gene Aliases: AA407365; AA986492; C1QBP; D11Wsu182e; gC1Q-R; GC1QBP; gC1qR; HABP1; P32; SF2P32
UniProt ID: (Human) Q07021, (Rat) O35796
Entrez Gene ID: (Human) 708, (Rat) 29681, (Mouse) 12261
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support