Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody tested in Indirect ELISA
Probable adipokine. Activates AMPK, AKT, and p44/42 MAPK signaling pathways
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complement C1q and tumor necrosis factor-related protein 9; Complement C1q and tumor necrosis factor-related protein 9A; complement C1q tumor necrosis factor-related protein 9; complement C1q tumor necrosis factor-related protein 9A
Gene Aliases: AQL1; C1QTNF9; C1QTNF9A; CTRP9; UNQ6503/PRO21380
UniProt ID: (Human) Q5VX66
Entrez Gene ID: (Human) 338872
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support