Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The epitope recognised by the antibody M81 is domain IV and/or V of the gamma-domain of activated C1s. Monoclonal antibody M81 blocks C4 activation and C4 binding to activated C1s. The antibody reacts around the binding site of C1s and reacts with both active and inactive C1s.
This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic proline-rich peptide IB-1; C1 esterase; Complement C1s subcomponent; Complement component 1 subcomponent s
Gene Aliases: C1S
UniProt ID: (Human) P09871
Entrez Gene ID: (Human) 716
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support