Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RNDKKDSQG RSNKALHLKS DAEFKKIFGL TKDLRVCLTR IPDHLTSGEG FDSFSSLVKS GTYKETEFMV KEGERKQQNF DKKRKAKTNK KMDHIKKRKT ENAYNAIING EANVTGSQLL SSILPTSDVS QHNILTSHSK TRQEKRTEME YYTHEKQEKG TLNSNAAYEQ SHFFNKNYTE DIFPVTPPEL EETIRDEKIR RLKQVLREKE AALEEMRKKM HQK (215-436 aa encoded by BC008115)
Together with SMCHD1, involved in chromosome X inactivation in females by promoting the compaction of heterochromatin (PubMed:23542155). Also able to repress the ligand-induced transcriptional activity of retinoic acid receptor alpha (RARA), possibly through direct recruitment of histone deacetylases (PubMed:17455211). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBiX1; HP1-binding protein enriched in inactive X chromosome protein 1; Ligand-dependent nuclear receptor-interacting factor 1; receptor interacting factor 1; Receptor-interacting factor 1
Gene Aliases: 2010012G17Rik; 4933421E11Rik; AI450568; C1orf103; LRIF1; RIF1
UniProt ID: (Human) Q5T3J3, (Mouse) Q8CDD9
Entrez Gene ID: (Human) 55791, (Mouse) 321000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support