Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: NGSTCKPDTE RQGTLSTAAP TTSPAPCLSN HHNK
Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 38%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein C1orf162
Gene Aliases: C1orf162
UniProt ID: (Human) Q8NEQ5
Entrez Gene ID: (Human) 128346
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support