Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ciliary microtubule-associated protein 2; Lymphocyte expansion molecule; uncharacterized protein C1orf177; uncharacterized protein C1orf177 homolog
Gene Aliases: C1orf177; CIMAP2; LEM; LEXM
UniProt ID: (Human) Q3ZCV2, (Rat) B1H283
Entrez Gene ID: (Human) 163747, (Rat) 500516
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support