Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSAWKTPRG SDAMPEIMVK IIGSKHFQYL VEKPKIKEND SLKTETQTMH QKPMTDNARQ MSRDTPVPIN FTDQQTTDNP DDVKEKKHPE NNQKSENNQK LLTGANSSRF LDGNIPSQAN VHCSSVPTGD QSLSYVHGIP RRKLRDWSLE QMVRGSSDQP EDIGQSPSGT TNEDAFLLAL VRRELKSRPL SSNLLEKLQK ELKILDPISS GFLLQSQLSR LFLKHEVPLQ LPTVKILCQR FSKRGSPEMV NYEKLLWFLN SAASDYPQQN KAAADLRKTE SHGTHSQSTP PQHSSSQPEV NRSLLEILKM ALRTTNGRLN IDNLNLSFRK EDRSFSGCLP LPKVRAICGK HGLYLTLSLL ETLLNHQDLG YQNEIKWQNF VEMLTRASSD LLSDLPTGKN EKKAPAPPME PEVPEMSQSK TEHMKTPEEE LQPESSPAET SACKDPLKPL KIRPVSQPFV NPAVKNKAEE CETWIDRFRK LENALYLCDL SNTGVLEKER ARRLIHNYNL IYNLSLSPQK IDQALRRFRS GENMLLEPAL RYLKEL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carcinoma-related EF-hand protein; Uncharacterized protein C1orf87
Gene Aliases: C1orf87; CREF
UniProt ID: (Human) Q8N0U7
Entrez Gene ID: (Human) 127795
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support