Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
MA1-40045 detects C1q from mouse samples.
MA1-40045 has been successfully used in immunohistochemistry (frozen), ELISA and immunofluorescence applications.
The MA1-40045 immunogen is mouse C1q component of the C1 complex.
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complement C1q subcomponent subunit A; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide
Gene Aliases: AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1qg; Ciqc
UniProt ID: (Mouse) P98086, (Mouse) P14106, (Mouse) Q02105
Entrez Gene ID: (Mouse) 12259, (Mouse) 12260, (Mouse) 12262
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support