Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRKRTEPVAL EHERCAAAGS SSSGSAAAAL DADCRLKQNL RLTGPAAAEP RCAADAGMKR ALGRRKGVWL RLRKILFCVL GLYIAIPFLI KLCPGIQAKL IFLNFVRVPY FIDLKKPQDQ GLNHTCNYYL QPEEDVTIGV WHTVPAVWWK NAQGKDQMWY EDALASSHPI ILYLHGNAGT RGGDHRVELY KVLSSLGYHV VTFDYRGWGD SVGTPSERGM TYDALHVFDW IKARSGDNPV YIWGHSLGTG VATNLVRRLC ERETPPDALI LESPFTNIRE EAKSHPFSVI YRYFPGFDWF FLDPITSSGI KFANDENVKH ISCPLLILHA EDDPVVPFQL GRKLYSIAAP ARSFRDFKVQ FVPFHSDLGY RHKYIYKSPE LPRILRPQQG PGSSPDPSMW SELV
This gene encodes an enzyme that catalyzes the hydrolysis of 2-arachidonoyl glycerol, the main endocannabinoid lipid transmitter that acts on cannabinoid receptors, CB1 and CB2. The endocannabinoid system is involved in a wide range of physiological processes, including neurotransmission, mood, appetite, pain appreciation, addiction behavior, and inflammation. Mutations in this gene are associated with the neurodegenerative disease, PHARC, resulting from an inborn error of endocannabinoid metabolism. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-arachidonoylglycerol hydrolase; 2-arachidonoylglycerol hydrolase ABHD12; Abhydrolase domain-containing protein 12; DKFZP434P106; hABHD12; Lysophosphatidylserine lipase ABHD12; Monoacylglycerol lipase ABHD12; Oxidized phosphatidylserine lipase ABHD12
Gene Aliases: ABHD12; ABHD12A; BEM46L2; C20orf22; dJ965G21.2; PHARC
UniProt ID: (Human) Q8N2K0
Entrez Gene ID: (Human) 26090
Molecular Function: serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support