Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: QETVMKFVPR YSLVLELSDS GAFRRSLHDP DGLVATYISE VHEHDGHLYL
Sequence homology: Cow: 77%; Dog: 92%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
APMAP (Adipocyte Plasma Membrane Associated Protein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include arylesterase activity and strictosidine synthase activity. Exhibits strong arylesterase activity with beta-naphthyl acetate and phenyl acetate. May play a role in adipocyte differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipocyte plasma membrane-associated protein; Protein BSCv
Gene Aliases: APMAP; BSCv; C20orf3; UNQ1869/PRO4305
UniProt ID: (Human) Q9HDC9
Entrez Gene ID: (Human) 57136
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support