Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). EDEM2 belongs to a family of proteins involved in ERAD of glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER degradation enhancer, mannosidase alpha-like 2; ER degradation-enhancing alpha-mannosidase-like 2; ER degradation-enhancing alpha-mannosidase-like protein 2; ER degradation-enhancing-mannosidase-like protein 2
Gene Aliases: 9530090G24Rik; AI327354; bA4204.1; C20orf31; C20orf49; EDEM2; RGD1303074; UNQ573/PRO1135
UniProt ID: (Human) Q9BV94, (Mouse) Q8BJT9
Entrez Gene ID: (Human) 55741, (Mouse) 108687, (Rat) 296304
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support