Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPFRLLIPLG LLCALLPQHH GAPGPDGSAP DPAHYRERVK AMFYHAYDSY LENAFPFDEL RPLTCDGHDT WGSFSLTLID ALDTLLILGN VSEFQRVVEV LQDSVDFDID VNASVFETNI RVVGGLLSAH LLSKKAGVEV EAGWPCSGPL LRMAEEAARK LLPAFQTPTG MPYGTVNLLH GVNPGETPVT CTAGIGTFIV EFATLSSLTG DPVFEDVARV ALMRLWESRS DIGLVGNHID VLTGKWVAQD AGIGAGVDSY FEYLVKGAIL LQDKKLMAMF LEYNKAIRNY TRFDDWYLWV QMYKGTVSMP VFQSLEAYWP GLQSLIGDID NAMRTFLNYY TVWKQFGGLP EFYNIPQGYT VEKREGYPLR PELIESAMYL YRATGDPTLL ELGRDAVESI EKISKVECGF ATIKDLRDHK LDNRMESFFL AETVKYLYLL FDPTNFIHNN GSTFDTVITP YGECILGAGG YIFNTEAHPI DPAALHCCQR LKEEQWEVED LMREFYSLKR SRSKFQKNTV SSGPWEPPAR PGTLFSPENH DQARERKPAK QKVPLLSCPS QPFTSKLALL GQVFLDSS
In the endoplasmic reticulum (ER), misfolded proteins are retrotranslocated to the cytosol and degraded by the proteasome in a process known as ER-associated degradation (ERAD). EDEM2 belongs to a family of proteins involved in ERAD of glycoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER degradation enhancer, mannosidase alpha-like 2; ER degradation-enhancing alpha-mannosidase-like 2; ER degradation-enhancing alpha-mannosidase-like protein 2; ER degradation-enhancing-mannosidase-like protein 2
Gene Aliases: bA4204.1; C20orf31; C20orf49; EDEM2; UNQ573/PRO1135
UniProt ID: (Human) Q9BV94
Entrez Gene ID: (Human) 55741
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support