Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKGTGIMDCA PKALLARALY DNCPDCSDEL AFSRGDILTI LEQHVPESEG WWKCLLHGRQ GLAPANRLQI LTEVAADRPC PPFLRGLEEA PASSEETYQV PTLPRPPTPG PVYEQMRSWA EGPQPPTAQV YEFPDPPTSA RIICEKTLSF PKQAILTLPR PVRASLPTLP SQVYDVPTQH RGPVVLKEPE KQQLYDIPAS PKKAGLHPPD SQASGQGVPL ISVTTLRRGG YSTLPNPQKS EWIYDTPVSP GKASVRNTPL TSFAEESRPH ALPSSSSTFY NPPSGRSRSL TPQLNNNVPM QKKLSLPEIP SYGFLVPRGT FPLDEDVSYK VPSSFLIPRV EQQNTKPNIY DIPKATSSVS QAGKELEKAK EVSENSAGHN SSWFSRRTTS PSPEPDRLSG SSSDSRASIV SSCSTTSTDD SSSSSSEESA KELSLDLDVA KETVMALQHK VVSSVAGLML FVSRKWRFRD YLEANIDAIH RSTDHIEESV REFLDFARGV HGTACNLTDS NLQNRIRDQM QTISNSYRIL LETKESLDNR NWPLEVLVTD SVQNSPDDLE RFVMVARMLP EDIKRFASIV IANGRLLFKR NCEKEETVQL TPNAEFKCEK YIQPPQRETE SHQKSTPSTK QREDEHSSEL LKKNRANICG QVSSEFQVIE KGASIVTWSS GY
Docking protein that plays a role in tyrosine kinase-based signaling related to cell adhesion and cell spreading. Regulates PTK2/FAK1 activity, focal adhesion integrity, and cell spreading. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cas scaffolding protein family member 4; HEF-like protein; HEF1-EFS-p130Cas-like protein; HEPL
Gene Aliases: C20orf32; CAS4; CASS4; HEFL; HEPL
UniProt ID: (Human) Q9NQ75
Entrez Gene ID: (Human) 57091
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support