Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMCPLWRLLI FLGLLALPLA PHKQPWPGLA QAHRDNKSTL ARIIAQGLIK HNAESRIQNI HFGDRLNASA QVAPGLVGWL ISGRKHQQQQ ESRSFDNNIV KMCAHMSIVV EFWLEKDEFG RRDLVIGKCD AEPSSVHVAI LTEAIPPKMN QFLYNLKENL QKVLPHMVES QVCPLIGEIL GQLDVKLLKS LIEQEAAHEP THHETSQPSA CQAGESPS
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPI fold containing family A, member 3; BPI fold-containing family A member 3; short long palate, lung and nasal epithelium carcinoma associated 3; Short palate, lung and nasal epithelium carcinoma-associated protein 3
Gene Aliases: BPIFA3; C20orf71; SPLUNC3
UniProt ID: (Human) Q9BQP9
Entrez Gene ID: (Human) 128861
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support