Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NLVQSVLSSR GVAQTPGSVE EDALLCGPVS KHKLPNQGED RRVPQNWFPI FNPERSDKPN ASDPSVPLKI PLQRNVIPSV TRVL
Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGME1; Mitochondrial genome maintenance exonuclease 1
Gene Aliases: bA504H3.4; C20orf72; DDK1; MGME1; MTDPS11
UniProt ID: (Human) Q9BQP7
Entrez Gene ID: (Human) 92667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support