Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
C2CD4D (C2 Calcium Dependent Domain Containing 4D) is a Protein Coding gene. An important paralog of this gene is C2CD4C.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2 calcium-dependent domain containing 4D; C2 calcium-dependent domain-containing protein 4D; family with sequence similarity 148, member D
Gene Aliases: C2CD4D; FAM148D; Gm659
UniProt ID: (Human) B7Z1M9, (Mouse) P0CG09
Entrez Gene ID: (Human) 100191040, (Rat) 100363111, (Mouse) 271944
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support