Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Glycosyltransferase that mediates core 2 O-glycan branching, an important step in mucin-type biosynthesis. Does not have core 4 O-glycan or I-branching enzyme activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-N-acetylg; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 4; C2GnT3; core 2 beta-1,6-N-acetylglucosaminyltransferase 3; Core 2-branching enzyme 3; Core2-GlcNAc-transferase 3; glucosaminyl (N-acetyl) transferase 3, mucin type pseudogene; glucosaminyl (N-acetyl) transferase 4, core 2 (beta-1,6-N-acetylglucosaminyltransferase)
Gene Aliases: C2GNT3; Gcnt4; Gm279; Gm73
UniProt ID: (Mouse) E9Q649
Entrez Gene ID: (Human) 51301, (Mouse) 218476
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support