Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The exact function of C2orf18 remains unknown. It contains 1 DUF6 domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANT2-binding protein; ANT2BP; CB018; Solute carrier family 35 member F6; solute carrier family 35, member F6; transmembrane protein C2orf18; Transport and Golgi organization 9 homolog
Gene Aliases: ANT2BP; C2orf18; SLC35F6; TANGO9; UNQ3047/PRO9863
UniProt ID: (Human) Q8N357
Entrez Gene ID: (Human) 54978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support