Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPPLLFEVS SLENAFQIGG HPWHYIVTPN KKKQKGVFHI CALKDNSLAK NGIQEMDCCS LESDWIYFHP DASGRIIHVG PNQVKVLKLT EIENNSSQHQ ISEDFVILAN RENHKNENVL TVTASGRVVK KSFNLLDDDP EQETFKIVDY EDELDLLSVV AVTQIDAEGK AHLDFHCNEY GTLLKSIPLV ESWDVTYSHE VYFDRDLVLH IEQKPNRVFS CYVYQMICDT GEEEETINRS C (281-520 aa encoded by BC140843)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDB1- and CUL4-associated factor 17
Gene Aliases: 2810055O12Rik; 4833418A01Rik; A030004A10Rik; A930009G19Rik; AI448937; C2orf37; DCAF17
UniProt ID: (Human) Q5H9S7, (Mouse) Q3TUL7
Entrez Gene ID: (Human) 80067, (Mouse) 75763
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support