Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Immunogen sequence: SHLLGKDKMS QMASVPEREP ESAPSAPSAE LQSTQHMEAQ PVESDADHVT AGANGQHGPQ AASTTKSAEE KAEHP
Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 31%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Testis-expressed protein 44
Gene Aliases: C2orf57; TEX44
UniProt ID: (Human) Q53QW1
Entrez Gene ID: (Human) 165100
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support