Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FAM228A (Family With Sequence Similarity 228 Member A) is a Protein Coding gene. Diseases associated with FAM228A include Echolalia. An important paralog of this gene is FAM228B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 228, member A; Protein FAM228A; UPF0638 protein C2orf84
Gene Aliases: C2orf84; FAM228A
UniProt ID: (Human) Q86W67
Entrez Gene ID: (Human) 653140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support