Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This antibody raised against 1314-1663 aa of human C3 protein recognize C3 alpha chain, C3b alpha' chain, and C3c alpha' chain fragment 2.
Immunogen sequence: ESASLLRSE ETKENEGFTV TAEGKGQGTL SVVTMYHAKA KDQLTCNKFD LKVTIKPAPE TEKRPQDAKN TMILEICTRY RGDQDATMSI LDISMMTGFA PDTDDLKQLA NGVDRYISKY ELDKAFSDRN TLIIYLDKVS HSEDDCLAFK VHQYFNVELI QPGAVKVYAY YNLEESCTRF YHPEKEDGKL NKLCRDELCR CAEENCFIQK SDDKVTLEER LDKACEPGVD YVYKTRLVKV QLSNDFDEYI MAIEQTIKSG SDEVQVGQQR TFISPIKCRE ALKLEEKKHY LMWGLSSDFW GEKPNLSYII GKDTWVEHWP EEDECQDEEN QKQCQDLGAF TESMVVFGCP N (1314-1663 aa encoded by BC150179)
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acylation-stimulating protein cleavage product; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a anaphylatoxin; Complement C3; complement component C3a; complement component C3b; epididymis secretory sperm binding protein Li 62p; prepro-C3
Gene Aliases: AHUS5; ARMD9; ASP; C3; C3a; C3b; CPAMD1; HEL-S-62p
UniProt ID: (Human) P01024
Entrez Gene ID: (Human) 718
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support