Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Specificity: Binds specifically to the 2e loop of the human C3a receptor, residues 185-193.
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; complement component 3 receptor 1
Gene Aliases: AZ3B; C3AR; C3AR1; C3R1; HNFAG09
UniProt ID: (Human) Q16581
Entrez Gene ID: (Human) 719
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support