Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: QSSWVTFRKI LSHFPEELSL AFVYGSGVYR QAGPSSDQKN AMLDFVFTVD
Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 93%; Horse: 92%; Human: 100%; Mouse: 77%; Pig: 100%; Rabbit: 86%; Rat: 93%
C3orf31 may be involved in the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDP-DAG synthase; CDP-diacylglycerol synthase; Mitochondrial translocator assembly and maintenance protein 41 homolog; MMP37-like protein, mitochondrial; Phosphatidate cytidylyltransferase, mitochondrial; TAM41; TAM41, mitochondrial translocator assembly and maintenance protein, homolog
Gene Aliases: C3orf31; RAM41; TAM41; TAMM41
UniProt ID: (Human) Q96BW9
Entrez Gene ID: (Human) 132001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support