Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DTVMEKRSFK VPLGKGRRCV VLADGFYEWQ RCQGTNQRQP YFIYFPQIKT EKSGSIGAAD SPENWEKVWD NWRLLTMAGI FDCWEPPEGG DVLYSYTIIT VDSCKGLSDI HHRMPAILDG EEAVSKWLDF GEVSTQEALK LIHPTENITF HAVSSVVNNS RNNTPECLAP VDLVVKKELR ASGSSQRMLQ WLATKSPKKE D
Sensor of abasic sites in single-stranded DNA (ssDNA) required to preserve genome integrity by promoting error-free repair of abasic sites (PubMed:30554877, PubMed:31235915, PubMed:31235913). Acts as an enzyme that recognizes and binds abasic sites in ssDNA at replication forks and chemically modifies the lesion by forming a covalent cross-link with DNA: forms a stable thiazolidine linkage between a ring-opened abasic site and the alpha-amino and sulfhydryl substituents of its N-terminal catalytic cysteine residue (PubMed:30554877, PubMed:31235913). The HMCES DNA-protein cross-link is then degraded by the proteasome (PubMed:30554877). Promotes error-free repair of abasic sites by acting as a 'suicide' enzyme that is degraded, thereby protecting abasic sites from translesion synthesis (TLS) polymerases and endonucleases that are error-prone and would generate mutations and double-strand breaks (PubMed:30554877). Has preference for ssDNA, but can also accommodate double-stranded DNA with 3' or 5' overhang (dsDNA), and dsDNA-ssDNA 3' junction (PubMed:31235915, PubMed:31806351). Also involved in class switch recombination (CSR) in B-cells independently of the formation of a DNA-protein cross-link: acts by binding and protecting ssDNA overhangs to promote DNA double-strand break repair through the microhomology-mediated alternative-end-joining (Alt-EJ) pathway. Acts as a protease: mediates autocatalytic processing of its N-terminal methionine in order to expose the catalytic cysteine. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abasic site processing protein HMCES; Embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; ES cell-specific 5hmC-binding protein; Peptidase HMCES; putative peptidase SRAPD1; SRAP domain-containing protein 1; UPF0361 protein C3orf37; UPF0361 protein C3orf37 homolog
Gene Aliases: 8430410A17Rik; C3orf37; C85376; DC12; HMCES; Srap1; SRAPD1
UniProt ID: (Human) Q96FZ2, (Mouse) Q8R1M0, (Rat) Q5XIJ1
Entrez Gene ID: (Human) 56941, (Mouse) 232210, (Rat) 500251
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support