Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
SMCO1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Single-pass membrane and coiled-coil domain-containing protein 1; Single-pass membrane protein with coiled-coil domains 1
Gene Aliases: C3orf43; SMCO1
UniProt ID: (Human) Q147U7
Entrez Gene ID: (Human) 255798
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support