Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQNEAPPNT HSIPGEPLYN YASIRLPEEH IPFFLHNNRH IATVCRKDSL CPYKKHLEKL KYCWGYEKSC KPEFRFGYPV CSYVDMGWTD TLESAEDIFW KQADFGYARE RLEEMHVLCQ PKETSDSSLV CSRYLQYCRA TNLYLDLRNI KRNHDRFKED FFQSGEIGGH CKLDIRTLTS EGQRKSPLQS WFAELQSYTQ LNFRPIEDAK CDIVIEKPTY FMKLDAGVNM YHHFCDFINL YITQHVNNSF STDVYIVMWD (18-276 aa encoded by BC060887)
The exact function of C3orf64 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AER61 glycosyltransferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF domain-specific O-linked N-acetylglucosamine transferase; EGF-O-GlcNAc transferase; Extracellular O-linked N-acetylglucosamine transferase
Gene Aliases: A130022J15Rik; AER61; AI447490; AOS4; AW214473; AW259391; C3orf64; EOGT; EOGT1
UniProt ID: (Human) Q5NDL2, (Mouse) Q8BYW9
Entrez Gene ID: (Human) 285203, (Mouse) 101351
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support