Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MLVGSQSFSP GGPNGIIRSQ SFAGFSGLQE RRSRCNSFIE NSSALKKPQA KLKKMHNLGH KNNNPPKEPQ PKRVEEVYRA LKNGLDEYLE VHQTELDKLT AQLKDMKRNS RLGVLYDLDK QIKTIERYMR RLEFHISKVD ELYEAYCIQR RLQDGASKMK QAFATSPASK AARESLTEIN RSFKEYTENM CTIEVELENL LGEFSIKMKG LAGFARLCPG DQYEIFMKYG RQRWKLKGKI EVNGKQSWDG EETVFLPLIV GFISIKVTEL KGLATHILVG SVTCETKELF AARPQVVAVD INDLGTIKLN LEITWYPFDV EDMTASSGAG NKAAALQRRM SMYSQGTPET PTFKDHSFFS NLPDDIFENG KAAEEKMPLS LSFSDLPNGD CALTSHSTGS PSNSTNPEIT ITPAEFNLSS LASQNEGMDD TSSASSRNSL GEGQEPKSHL KEEDPEEPRK PASAPSEACR RQSSGAGAEH LFLENDVAEA LLQESEEASE LKPVELDTSE GNITKQLVKR LTSAEVPMAT DRLLSEGSVG GESEGCRSFL DGSLEDAFNG LLLALEPHKE QYKEFQDLNQ EVMNLDDILK K
Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 65, member B; myogenesis-related and NCAM-associated protein homolog; Rho family-interacting cell polarization regulator 2
Gene Aliases: C6orf32; DFNB104; DIFF40; DIFF48; FAM65B; KIAA0386; MYONAP; PL48; RIPOR2
UniProt ID: (Human) Q9Y4F9
Entrez Gene ID: (Human) 9750
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support