Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGKANASKK NAQQLKRNPK RKKDNEEVVL SENKVRNTVK KNKNHLKDLS SEGQTKHTNL KHGKTAASKR KTWQPLSKST RDHLQTMMES VIMTILSNSI KEKEEIQYHL NFLKKRLLQQ CETLKVPPKK MEDLTNVSSL LNMERARDKA NEEGLALLQE EIDKMVETTE LMTGNIQSLK NKIQILASEV EEEEERVKQM HQINSSGVLS LPELSQKTLK APTLQKEILA LIPNQNALLK DLDILHNSSQ MKSMSTFIEE AYKKLDAS
CENPQ is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP-Q; Centromere protein Q
Gene Aliases: C6orf139; CENP-Q; CENPQ
UniProt ID: (Human) Q7L2Z9
Entrez Gene ID: (Human) 55166
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support