Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
METTL24 gene ontology annotations related to this gene include methyltransferase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: methyltransferase-like protein 24; Probable methyltransferase-like protein 24; UPF0624 protein C6orf186; UPF0624 protein C6orf186 homolog
Gene Aliases: 9030224M15Rik; C6orf186; dJ71D21.2; METTL24
UniProt ID: (Human) Q5JXM2, (Mouse) Q8CCB5, (Rat) Q5BK01
Entrez Gene ID: (Human) 728464, (Mouse) 327747, (Rat) 499465
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support