Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LEAITTASEN IGKMVASLQS DIIESQACQD MQNEQKVYAK IQATHKQREL ANKSWEDFLA RTSNAKTLKK AKRLQEKAIE ASRYHKRPQN AIFHQTHIKG LNTMVPSGGV VTVESTPARL VGGPLVDTDI ALKKLPIRGG ALQRVKAVKI VDAPKKSIPT
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 69
Gene Aliases: 4921525K03; A330021E22Rik; AI427898; Cfap69
UniProt ID: (Mouse) Q8BH53
Entrez Gene ID: (Mouse) 207686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support