Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 69; uncharacterized LOC100910867
Gene Aliases: 4921525K03; A330021E22Rik; AI427898; C7orf63; CFAP69; FAP69
UniProt ID: (Human) A5D8W1, (Mouse) Q8BH53
Entrez Gene ID: (Human) 79846, (Mouse) 207686, (Rat) 100910867
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support