Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: SGLKKIFKLS AAVVLIGSHP NLSFLKDQGC YLGHKSSQPI TCKGNPVEID TYTYECIKEA NLFALGPLVG DNFVRFLKGG ALGVTRCLAT RQKKKHLFVE RGGGDGIA
May be involved in meiosis or the maturation of germ cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hT41; Oxidative stress-induced growth inhibitor 2
Gene Aliases: C8orf1; hT41; OSGIN2
UniProt ID: (Human) Q9Y236
Entrez Gene ID: (Human) 734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support