Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPLVEETSLL EDSSVTFPVV IIGNGPSGIC LSYMLSGYRP YLSSEAIHPN TILNSKLEEA RHLSIVDQDL EYLSEGLEGR SSNPVAVLFD TLLHPDADFG YDYPSVLHWK LEQHHYIPHV VLGKGPPGGA WHNMEGSMLT ISFGSWMELP GLKFKDWVSS KRRSLKGDRV MPEEIARYYK HYVKVMGLQK NFRENTYITS VSRLYRDQDD DDIQDRDIST KHLQIEKSNF IKRNWEIRGY QRIADGSHVP FCLFAENVAL ATGTLDSPAH LEIEGEDFPF VFHSMPEFGA AINKGKLRGK VDPVLIVGSG LTAADAVLCA YNSNIPVIHV FRRRVTDPSL IFKQLPKKLY PEYHKVYHMM CTQSYSVDSN LLSDYTSFPE HRVLSFKSDM KCVLQSVSGL KKIFKLSAAV VLIGSHPNLS FLKDQGCYLG HKSSQPITCK GNPVEIDTYT YECIKEANLF ALGPLVGDNF VRFLKGGALG VTRCLATRQK KKHLFVERGG GDGIA
May be involved in meiosis or the maturation of germ cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hT41; Oxidative stress-induced growth inhibitor 2
Gene Aliases: C8orf1; hT41; OSGIN2
UniProt ID: (Human) Q9Y236
Entrez Gene ID: (Human) 734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support