Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: KRELATGRK AMQRLAKKMS GWFLRRTKTL IKDQLPKKED RMVYCSLTDF QKAVYQTVLE TEDVTLILQS SEPCTCRSGQ KRRNCCYKTN SHGETVKTLY LSYLTVLQKV ANHVALLQAA STSKQQETLI KRICDQVFSR FPDFVQKSKD AAFETLSDPK YSGKMKVLQQ LLNHCRKNRD KVLLFSFSTK LLDVLQQYCM ASGLDYRRLD GSTKSEERLK IVKEFNSTQD VNICLVSTMA GGLGLNFVGA NVVVLFDPTW NPANDLQAID RAYRIGQCRD VKVLRLISLG TVEEIMYLRQ IYKQQLHCVV VGSENAKRYF EAVQGSKEHQ GELFGIHNLF KFRSQGSCLT KDILEV (358-712 aa encoded by BC140702)
This gene encodes a member of the Snf2 family of helicase-like proteins. The encoded protein may play a role in DNA repair and mitochondrial function. Mutations in this gene have been associated with bone marrow failure syndrome 2. Alternatively spliced transcript variants that encode different protein isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA excision repair protein ERCC-6-like 2; DNA repair and recombination protein RAD26-like; excision repair cross-complementation group 6 like 2; excision repair cross-complementation group 6-like 2; excision repair cross-complementing rodent repair deficiency, complementation group 6-like 2; putative repair and recombination helicase RAD26L; stretch responsive protein 278
Gene Aliases: BMFS2; C9orf102; ERCC6L2; RAD26L; SR278
UniProt ID: (Human) Q5T890
Entrez Gene ID: (Human) 375748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support