Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amplimer WI-13707, mouse homolog; ci150; leucine repeat adaptor protein 35b; Leucine rich adaptor protein 1-like
Gene Aliases: 1110029A09Rik; AV077978; AV175137; bA3L8.2; bM350F23.1; C9orf150; D4Bwg0951e; HYST0841; LRAP35b; LURAP1L; RGD1308059; WI-13707
UniProt ID: (Human) Q8IV03, (Rat) Q5BJW5, (Mouse) Q8K2P1
Entrez Gene ID: (Human) 286343, (Rat) 362535, (Mouse) 52829
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support