Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The protein encoded by this gene has homologs that have been identified in mouse, macaque, etc organisms. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 219, member A; likely ortholog of H. sapiens chromosome 9 open reading frame 25 (C9orf25); Protein FAM219A; uncharacterized protein C9orf25; uncharacterized protein C9orf25 homolog
Gene Aliases: 2310028H24Rik; C9orf25; FAM219A
UniProt ID: (Human) Q8IW50, (Mouse) Q9D772
Entrez Gene ID: (Human) 203259, (Mouse) 71901, (Rat) 691024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support