Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDIQLDPARD DLPLMANTSH ILVKHYVLDL DVDFESQVIE GTIVLFLEDG NRFKKQNSSI EEACQSESNK ACKFGMPEPC HIPVTNARTF SSEMEYNDFA ICSKGEKDTS DKDGNHDNQE HASGISSSKY CCDTGNHGSE DFLLVLDCCD LSVLKVEEVD VAAVPGLEKF TRSPELTVVS EEFRNQIVRE LVTLPANRWR EQLDYYARCS QAPGCGELLF DTDTWSLQIR KTGAQTATDF PHAIRIWYKT KPEGRSVTWT SDQSGRPCVY TVGSPINNRA LFPCQEPPVA MSTWQATVRA AASFVVLMSG ENSAKPTQLW EECSSWYYYV TMPMPASTFT IAVGCWTEMK METWSSNDLA TERPFSPSEA NFRHVGVCSH MEYPCRFQNA SATTQEIIPH RVFAPVCLTG ACQETLLRLI PPCLSAAHSV LGAHPFSRLD VLIVPANFPS LGMARPSKDK TGHTSDSGAS VIKHGLNPEK IFMQVHYLKG YFLLRFLAKR LGDETYFSFL RKFVHTFHGQ LILSQDFLQM LLENIPEEKR LELSVENIYQ DWLESSGIPK PLQRERRAGA ECGLARQVRA EVTKWIGVNR RPRKRKRREK EEVFEKLLPD QLVLLLEHLL EQKTLSPRTL QSLQRTYHLQ DQDAEVRHRW CELIVKHKFT KAYKSVERFL QEDQAMGVYL YGELMVSEDA RQQQLARRCF ERTKEQMDRS SAQVVAEMLF
Aminopeptidases catalyze the hydrolysis of amino acid residues from the N-terminus of peptide or protein substrates. Able to cleave angiotensin III to generate angiotensin IV, a bioactive peptide of the renin-angiotensin pathway. Not able to cleave angiotensin I and angiotensin II. May play a role in the proteolytic processing of bioactive peptides in tissues such as testis and heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminopeptidase O; AP-O; AP-OFLJ55832; FLJ14675; FLJ40923
Gene Aliases: AOPEP; AP-O; APO; C90RF3; C9orf3; ONPEP
UniProt ID: (Human) Q8N6M6
Entrez Gene ID: (Human) 84909
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support