Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spermatogenesis associated 6-like protein
Gene Aliases: bA6J24.2; C9orf68; SPATA6L
UniProt ID: (Human) Q8N4H0
Entrez Gene ID: (Human) 55064
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support