Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 11
Immunogen sequence: SPAVAKTEIA LSGKSPLLAA TFAYWDNILG PRVRHIWAPK TEQVLLSDGE ITFLANHTLN GEILRNAESG AIDVKFFVLS EKGVIIVSLI FDGNWNGDRS TYGLSIILPQ TELSFYLPLH RVCVDRLTHI IRKGRIWMHK ERQENVQKII LEGTERMEDQ GQSIIPMLTG EVIPVMELLS SMKSHSVPEE ID
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Guanine nucleotide exchange factor C9orf72; Guanine nucleotide exchange factor C9orf72 homolog; protein C9orf72 homolog
Gene Aliases: AI840585; ALSFTD; C9orf72; DENND9; DENNL72; FTDALS; FTDALS1
UniProt ID: (Mouse) Q6DFW0, (Human) Q96LT7, (Rat) Q66HC3
Entrez Gene ID: (Mouse) 73205, (Human) 203228, (Rat) 313155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support