Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: PPFLPAASEE AEPAEGLRVP GLAKNSREYV RPGLPVTFID EVDSEEAPQA
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%
The specific function of C9orf75 is not yet known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Taperin
Gene Aliases: C9orf75; DFNB79; TPRN
UniProt ID: (Human) Q4KMQ1
Entrez Gene ID: (Human) 286262
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support