Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAM75-like protein C9orf79; family with sequence similarity 75, member E1; Protein FAM75E1; SPATA31 subfamily E, member 1; Spermatogenesis-associated protein 31E1; XXyac-YM21GA2.5
Gene Aliases: C9orf79; FAM75E1; SPATA31E1
UniProt ID: (Human) Q6ZUB1
Entrez Gene ID: (Human) 286234
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support