Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: PFDSIMERLT LRRIDPVTGE RYHLMYKPPP TMEIQARLLQ NPKDAEEQVK LKMDLFYRNS ADLEQLYGSA ITLNGDQDPY TVFEYIESGI INPLPKKIP
Enables AMP binding activity and nucleobase-containing compound kinase activity. Involved in nucleoside diphosphate phosphorylation and nucleoside triphosphate biosynthetic process. Located in 9+2 motile cilium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase 8; AK 8; ATP-AMP transphosphorylase 8; putative adenylate kinase-like protein C9orf98; RP11-143F18.1
Gene Aliases: AK 8; AK8; C9orf98; DDX31
UniProt ID: (Human) Q96MA6
Entrez Gene ID: (Human) 158067
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support