Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MASPDWGYD DKNGPEQWSK LYPIANGNNQ SPVDIKTSET KHDTSLKPIS VSYNPATAKE IINVGHSFHV NFEDNDNRSV LKGGPFSDSY RLFQFHFHWG STNEHGSEHT VDGVKYSAEL HVAHWNSAKY SSLAEAASKA DGLAVIGVLM KVGEANPKLQ KVLDALQAIK TKGKRAPFTN FDPSTLLPSS LDFWTYPGSL THPPLYESVT WIICKESISV SSEQLAQFRS LLSNVEGDNA VPMQHNNRPT QPLKGRTVRA SF (1-261 aa encoded by BC027890)
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA1 is closely linked to CA2 and CA3 genes on chromosome 8, and it encodes a cytosolic protein which is found at the highest level in erythrocytes. Variants of this gene have been described in some populations. Multiple alternatively spliced variants, encoding the same protein, have been identified. Transcript variants of CA1 utilizing alternative polyA_sites have been described in literature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA I; CA-I; CA-I {ECO:0000250|UniProtKB:P00915}; ca1 {ECO:0000250|UniProtKB:P00915}; CAB; Carbonate dehydratase I; carbonate dehydratase I {ECO:0000250|UniProtKB:P00915}; Carbonic anhydrase 1; carbonic anhydrase 1 {ECO:0000250|UniProtKB:P00915}; Carbonic anhydrase B; Carbonic anhydrase I; carbonic anhydrase I {ECO:0000250|UniProtKB:P00915}; Cyanamide hydratase CA1; epididymis secretory protein Li 11
Gene Aliases: AW555628; CA-I; CA1; CAB; Car-1; Car1; HEL-S-11
UniProt ID: (Human) P00915, (Rat) B0BNN3, (Mouse) P13634
Entrez Gene ID: (Human) 759, (Rat) 310218, (Mouse) 12346
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support