Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: MSHHWGYGK HNGPEHWHKD FPIAKGERQS PVDIDTHTAK YDPSLKPLSV SYDQATSLRI LNNGHAFNVE FDDSQDKAVL KGGPLDGTYR LIQFHFHWGS LDGQGSEHTV DKKKYAAELH LVHWNTKYGD FGKAVQQPDG LAVLGIFLKV GSAKPGLQKV VDVLDSIKTK GKSADFTNFA ARGLLPESLD YWTYPGSLTT PPLLECVTWI VLKEPISVSS EQVLKFRKLN FNGEGEPEEL MVDNWRPAQP LKNRQIKASF K (1-260 aa encoded by BC011949)
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA II; CA-II; CAC; Carbonate dehydratase II; Carbonic anhydrase 2; carbonic anhydrase B; Carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; Cyanamide hydratase CA2; epididymis luminal protein 76; epididymis secretory protein Li 282
Gene Aliases: AI131712; CA-II; CA2; CAC; CAII; Car-2; Car2; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
UniProt ID: (Human) Q6FI12, (Mouse) P00920, (Rat) P27139
Entrez Gene ID: (Human) 760, (Mouse) 12349, (Rat) 54231
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support