Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LQHYYTYHGS LTTPPCTENV HWFVLADFVK LSRTQVWKLE NSLLDHRNKT IHNDYRRTQP LNHRVVESNF PNQEYTLGSE FQFYLHKIEE ILDYLRRALN
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA-6; CA-VI; Carbonate dehydratase VI; Carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; Salivary carbonic anhydrase; Secreted carbonic anhydrase
Gene Aliases: CA-VI; CA6; GUSTIN
UniProt ID: (Human) P23280
Entrez Gene ID: (Human) 765
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support