Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Carbonic Anhydrase IX Antibodies

            Proteintech

            CA9 Polyclonal Antibody

            40 References
            View all (67) Carbonic Anhydrase IX antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite CA9 Polyclonal Antibody

            • Antibody Testing Data (30)
            Application
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 30

            CA9 Antibody (11071-1-AP) in IHC (P)

            Immunohistochemistry of paraffin-embedded human renal cell carcinoma tissue slide using 11071-1-AP ( CA IX antibody at dilution of 1:50 (under 40x lens).
            View Product

            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Western Blot (WB)
            CA9 Antibody in Flow Cytometry (Flow)
            CA9 Antibody in Immunoprecipitation (IP)

            Product Details

            11071-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            View 15 publications 15 publications

            Immunohistochemistry (IHC)

            -
            View 22 publications 22 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            -
            View 3 publications 3 publications

            Flow Cytometry (Flow)

            0.2 µg/1x10^6 cells
            -

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            CA9 Fusion Protein Ag1540 (31-408 aa encoded by BC014950)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.27 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: LLMPVHPQR LPRMQEDSPL GGGSSGEDDP LGEEDLPSEE DSPREEDPPG EEDLPGEEDL PGEEDLPEVK PKSEEEGSLK LEDLPTVEAP GDPQEPQNNA HRDKEGDDQS HWRYGGDPPW PRVSPACAGR FQSPVDIRPQ LAAFCPALRP LELLGFQLPP LPELRLRNNG HSVQLTLPPG LEMALGPGRE YRALQLHLHW GAAGRPGSEH TVEGHRFPAE IHVVHLSTAF ARVDEALGRP GGLAVLAAFL EEGPEENSAY EQLLSRLEEI AEEGSETQVP GLDISALLPS DFSRYFQYEG SLTTPPCAQG VIWTVFNQTV MLSAKQLHTL SDTLWGPGDS RLQLNFRATQ PLNGRVIEAS FPAGVDSSPR AAEPVQLNS (31-408 aa encoded by BC014950)

            Target Information

            Carbonic anhydrase (CA) is an enzyme that assists rapid interconversion of carbon dioxide and water into carbonic acid, protons, and bicarbonate ions. It is abundant in all mammalian tissues. There are many genes that are inducible by hypoxia, via HIF-1 alpha. CA IX is one of the most inducible genes because of its stability and location within the membrane. Carbonic anhydrases have a widespread role in regulating pH in normal tissues, by regulating hydrogen ion (H+) flux. The pH is important in cell death under hypoxia, thus a blockade of CA IX results in increased cell death under hypoxia. Therefore, CA IX has become a reliable histochemical marker of hypoxia.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: CA-9; CA-IX; Carbonate dehydratase IX; Carbonic anhydrase 9; Carbonic anhydrase IX; carbonic dehydratase; Membrane antigen MN; Membrane antigen MN homolog; P54 / 58N; P54/58N; pMW1; RCC-associated antigen G250; RCC-associated protein G250; Renal cell carcinoma-associated antigen G250

            View more View less

            Gene Aliases: CA9; CAIX; Car9; G250; MN; MN/CA9

            View more View less

            UniProt ID: (Human) Q16790, (Mouse) Q8VHB5

            View more View less

            Entrez Gene ID: (Human) 768, (Mouse) 230099, (Rat) 313495

            View more View less

            Function(s)

            carbonate dehydratase activity zinc ion binding lyase activity metal ion binding molecular_function

            Process(es)

            morphogenesis of an epithelium one-carbon metabolic process secretion response to hypoxia response to testosterone response to drug bicarbonate transport regulation of transcription from RNA polymerase II promoter in response to hypoxia
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-fsh6n:80/100.66.78.219:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0