Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MTEPWLALGT SWTLPLQDRE LGPEELDELQ AAFEEFDTDR DGYISHRELG DCMRTLGYMP TEMELLEVSQ HIKMRMGGRV DFEEFVELIG PKLREETAHM LGVRELRIAF REFDRDRDGR ITVAELREAV PALLGEPLAG PELDEMLREV DLNGDGTVDF DEFVMMLSRH
The calcium binding protein (CaBP) family shares much similarity to calmodulin. It has been shown that CaBP proteins can substitute functionally for, and probably augment the function of, calmodulin. Calcium binding proteins are a crucial part of calcium mediated cellular signal transduction in the central nervous system. There are several members of the family with varying expression patterns. CaBP1 and CaBP2 can be expressed as multiple, alternatively spliced variants in brain and retina. CaBP3, CaBP4 and CaBP 5 are restricted to retinal rod and cone cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CaBP4; Calcium-binding protein 4
Gene Aliases: CABP4; CRSD; CSNB2B
UniProt ID: (Human) P57796
Entrez Gene ID: (Human) 57010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support