Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is Hela cell lysate.
PA5-21001 can be used with blocking peptide PEP-1115.
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CaBP7; Calcium-binding protein 7; calcium-binding protein CaBP7; calneuron 2; Calneuron II; Calneuron-2; calneuron-II
Gene Aliases: CABP7; CALN2; rCaBP7
UniProt ID: (Human) Q86V35, (Mouse) Q91ZM8, (Rat) Q66H96
Entrez Gene ID: (Human) 164633, (Mouse) 192650, (Rat) 360970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support